3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals
3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals
3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals
3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number ACR294390100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery white
FormSlug, 6.35mm (0.25in) diameter.
Assay from Supplier's CofA≥99.99% (metals basis)