View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 6-Di-tert-butyl-4-methoxyphenol
Name Note97%
CAS489-01-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning