2-Bromoacetamide, 98%, Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
CAS: 683-57-8 | C2H4BrNO | 137.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR291100250
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2868-37-3
IUPAC Namemethyl cyclopropanecarboxylate
Molecular FormulaC5H8O2
InChI KeyPKAHQJNJPDVTDP-UHFFFAOYSA-N
SMILESCOC(=O)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4165-1.4205 @ 20?C