Selenophene, 99%, Thermo Scientific Chemicals
Selenophene, 99%, Thermo Scientific Chemicals
Selenophene, 99%, Thermo Scientific Chemicals
Selenophene, 99%, Thermo Scientific Chemicals
Acros Organics
Selenophene, 99%, Thermo Scientific Chemicals
CAS: 288-05-1 | C4H4Se | 131.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number ACR271900100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9005-38-3
Molecular Formula(C6H7O7)A(C6H7O7)BNa
SMILES[Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O
Molecular Weight (g/mol)NaN
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or yellowish brown
Loss on Drying≤13.0%
Total ash18-27% (dried basis)
pH5.0-7.5 (1% solution)
Viscosity30-90 mPas (1% aq. soln.)