2,2,4-Trimethylpentane, 99.5%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, Thermo Scientific Chemicals
For HPLC
Have Questions?
Catalog NumberQuantityPackaging
ACR2688100101 LGlass Bottle
Catalog number ACR268810010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS12068-85-8
IUPAC Namedisulfanylideneiron
Molecular FormulaFeS2
InChI KeyNFMAZVUSKIJEIH-UHFFFAOYSA-N
SMILESS=[Fe]=S
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
For HPLC
RUO – Research Use Only