3-Amino-1H-1,2,4-triazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-1,2,4-triazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-1,2,4-triazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-1,2,4-triazole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Amino-1H-1,2,4-triazole, 95%, Thermo Scientific Chemicals
CAS: 61-82-5 | C2H4N4 | 84.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number ACR264570100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark grey to black
Elemental Analysis4.80 to 5.10 Pd wt% ( dry basis)
Formpowder
Loss on Drying40.0 to 65.0 wt%