Buffer solution pH 5, Acetate buffer
Buffer solution pH 5, Acetate buffer
Buffer solution pH 5, Acetate buffer
Buffer solution pH 5, Acetate buffer
Thermo Scientific Chemicals
Buffer solution pH 5, Acetate buffer
CAS: 64-19-7
Have Questions?
Catalog NumberQuantity
ACR2586200101 L
Catalog number ACR258620010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver to silver-grey
Assay (unspecified)Indium (In): ≥99.99% (metals basis)
FormFoil
pH buffer
RUO – Research Use Only