Sodium, 30wt% dispersion in toluene, <0.1 mm particle size
Sodium, 30wt% dispersion in toluene, &lt;0.1 mm particle size
Sodium, 30wt% dispersion in toluene, &lt;0.1 mm particle size
Sodium, 30wt% dispersion in toluene, &lt;0.1 mm particle size
Thermo Scientific Chemicals
Sodium, 30wt% dispersion in toluene, <0.1 mm particle size
CAS: 7440-23-5 | Na | 22.99 g/mol
Have Questions?
Catalog NumberQuantity
ACR243262500250 mL
Catalog number ACR243262500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS89763-93-9
IUPAC Name2-fluoro-4-(trifluoromethyl)benzaldehyde
Molecular FormulaC8H4F4O
InChI KeyKFEHNXLFIGPWNB-UHFFFAOYSA-N
SMILESFC1=CC(=CC=C1C=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Free acid (titration)≤3.0%
Refractive Index1.4475-1.4525 @ 20?C