Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
CAS: 108-73-6 | C6H6O3 | 126.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number ACR241761000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS93-60-7
IUPAC Namemethyl pyridine-3-carboxylate
Molecular FormulaC7H7NO2
InChI KeyYNBADRVTZLEFNH-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point38°C to 43°C
Titration with HClO4>=98.5 %
Appearance (Color)White to brown