Silica gel, for column chromatography, 0.200-0.500mm, 40Å
Silica gel, for column chromatography, 0.200-0.500mm, 40Å
Silica gel, for column chromatography, 0.200-0.500mm, 40Å
Silica gel, for column chromatography, 0.200-0.500mm, 40Å
Silica gel, for column chromatography, 0.200-0.500mm, 40Å
Thermo Scientific Chemicals
Silica gel, for column chromatography, 0.200-0.500mm, 40Å
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number ACR241672500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Pore Size40 Å
PackagingPlastic Bottle
Quantity250 g
TypeSilica Gel
MaterialSilica Gel
View more
For column chromatography
RUO – Research Use Only