L-alpha-Dipalmitoyl phosphatidylcholine, 98%, Thermo Scientific Chemicals
L-alpha-Dipalmitoyl phosphatidylcholine, 98%, Thermo Scientific Chemicals
L-alpha-Dipalmitoyl phosphatidylcholine, 98%, Thermo Scientific Chemicals
L-alpha-Dipalmitoyl phosphatidylcholine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L-alpha-Dipalmitoyl phosphatidylcholine, 98%, Thermo Scientific Chemicals
CAS: 63-89-8 | C40H80NO8P | 734.053 g/mol
Have Questions?
Catalog NumberQuantity
ACR240571000100 mg
Catalog number ACR240571000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialL-α-Dipalmitoyl phosphatidylcholine
Name Note98%
CAS63-89-8
Health Hazard 1HYGROSCOPIC