Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
Packaging:
View all
Catalog number ACR232110010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS124-41-4
IUPAC Namesodium methanolate
Molecular FormulaCH3NaO
InChI KeyWQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES[Na+].C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Solution
Strength5.3 to 5.6 M
For analysis
RUO – Research Use Only