D(+)-Biotin, 98%
D(+)-Biotin, 98%
D(+)-Biotin, 98%
Thermo Scientific Chemicals
D(+)-Biotin, 98%
CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number ACR230095000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD(+)-Biotin
CAS58-85-5
Melting Point231.0°C to 233.0°C
EINECS Number200-399-3
Assay Percent Range98%