D(+)-Biotin, 98%
D(+)-Biotin, 98%
D(+)-Biotin, 98%
Thermo Scientific Chemicals
D(+)-Biotin, 98%
CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 mg
1 g
25 g
Packaging:
View all
Glass bottle
Catalog number ACR230090050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106-70-7
IUPAC Namemethyl hexanoate
Molecular FormulaC7H14O2
InChI KeyNUKZAGXMHTUAFE-UHFFFAOYSA-N
SMILESCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Refractive Index1.4035-1.4065 @ 20?C