γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
CAS: 91464-90-3 | C48H80O40 | 1297.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR229905000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS24057-28-1
IUPAC Name4-methylbenzene-1-sulfonic acid; pyridine
Molecular FormulaC12H13NO3S
InChI KeyZDYVRSLAEXCVBX-UHFFFAOYSA-N
SMILESC1=CC=NC=C1.CC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Melting point112°C to 119°C
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Chelation/Complexation reagent
RUO – Research Use Only