Potassium antimonyl tartrate trihydrate, 99+%, Thermo Scientific Chemicals
Potassium antimonyl tartrate trihydrate, 99+%, Thermo Scientific Chemicals
Potassium antimonyl tartrate trihydrate, 99+%, Thermo Scientific Chemicals
Potassium antimonyl tartrate trihydrate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium antimonyl tartrate trihydrate, 99+%, Thermo Scientific Chemicals
CAS: 28300-74-5 | 1·5 H2O | 333.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR223805000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4805-22-5
IUPAC Name5,5'-dibromo-2,2'-bithiophene
Molecular FormulaC8H4Br2S2
InChI KeySXNCMLQAQIGJDO-UHFFFAOYSA-N
SMILESBrC1=CC=C(S1)C1=CC=C(Br)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥97.5%