Lead, +99%, granular, Thermo Scientific Chemicals
Lead, +99%, granular, Thermo Scientific Chemicals
Lead, +99%, granular, Thermo Scientific Chemicals
Lead, +99%, granular, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lead, +99%, granular, Thermo Scientific Chemicals
CAS: 7439-92-1 | Pb | 207.20 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR222621000100 gPlastic bottle
Catalog number ACR222621000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialLead
ColorGray
Name Notep.a.
CAS7439-92-1
Melting Point327.4°C