Phenol, 99.5%, extra pure, loose crystals, unstabilized, Thermo Scientific Chemicals
Phenol, 99.5%, extra pure, loose crystals, unstabilized, Thermo Scientific Chemicals
Phenol, 99.5%, extra pure, loose crystals, unstabilized, Thermo Scientific Chemicals
Phenol, 99.5%, extra pure, loose crystals, unstabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenol, 99.5%, extra pure, loose crystals, unstabilized, Thermo Scientific Chemicals
CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Packaging:
Glass bottle
Catalog number ACR221752500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenol
Name Notep.a.
Acidity(per Eur. Pharm.) Passes Test
CAS108-95-2
Health Hazard 1GHS Signal Word: Danger