Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 62-56-6 | CH4N2S | 76.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR220050010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS20260-53-1
IUPAC Namehydrogen pyridine-3-carbonyl chloride chloride
Molecular FormulaC6H5Cl2NO
InChI KeyMSYBLBLAMDYKKZ-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClC(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to gray
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥95.0 to ≤105.0%
For analysis
RUO – Research Use Only