Allyl methacrylate, 98%, stabilized, Thermo Scientific Chemicals
Allyl methacrylate, 98%, stabilized, Thermo Scientific Chemicals
Allyl methacrylate, 98%, stabilized, Thermo Scientific Chemicals
Allyl methacrylate, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allyl methacrylate, 98%, stabilized, Thermo Scientific Chemicals
CAS: 96-05-9 | C7H10O2 | 126.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR215875000500 mLGlass bottle
Catalog number ACR215875000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAllyl methacrylate
Acidity0.05% max.
CAS96-05-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Fatal if inhaled.
Toxic in contact with skin.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure in contact with skin.
Flammable liquid and vapor.
Very toxic to aqu