Ammonium cerium(IV) nitrate, pure, Thermo Scientific Chemicals
Ammonium cerium(IV) nitrate, pure, Thermo Scientific Chemicals
Ammonium cerium(IV) nitrate, pure, Thermo Scientific Chemicals
Ammonium cerium(IV) nitrate, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium cerium(IV) nitrate, pure, Thermo Scientific Chemicals
Oxidation reagent
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number ACR212360010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium cerium(IV) nitrate
Melting Point107.0°C to 108.0°C
CAS16774-21-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.