N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
500 mL
1 L
2.5 L
25 L
Packaging:
View all
Plastic Drum
Catalog number ACR210580100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS1772-03-8
IUPAC Namehydrogen (3R,4R,5R,6R)-3-amino-6-(hydroxymethyl)oxane-2,4,5-triol chloride
Molecular FormulaC6H14ClNO5
InChI KeyQKPLRMLTKYXDST-BMZZJELJSA-N
SMILES[H+].[Cl-].N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 % (K.F.)
Specific optical rotation+93° to +99° (20°C, 589 nm) (c=1, H2O)
Specific optical rotationafter 24 h
Residue after ignition=<1 %
Appearance (Color)White to off-white
View more
For analysis
RUO – Research Use Only