Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
CAS: 12501-23-4 | H5O41PW12 | 2880.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR208311000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS2564-83-2
IUPAC Name(2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Molecular FormulaC9H18NO
InChI KeyQYTDEUPAUMOIOP-UHFFFAOYSA-N
SMILESCC1(C)CCCC(C)(C)N1[O]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange-red to red to dark red
FormCrystalline powder or powder or crystals or fused solid and/or chunks
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)31-40°C