CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
Thermo Scientific Chemicals
CHES, 98+%, for biochemistry
CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number ACR208181000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS2867-47-2
IUPAC Name2-(dimethylamino)ethyl 2-methylprop-2-enoate
Molecular FormulaC8H15NO2
InChI KeyJKNCOURZONDCGV-UHFFFAOYSA-N
SMILESCN(C)CCOC(=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.4390 to 1.4410 (20°C, 589 nm)
Appearance (Form)Liquid
Stabilizer750 to 2000 ppm MEHQ
Appearance (Color)Clear colorless to yellow
View more
Biological buffer
RUO – Research Use Only