Spironolactone, 99%
Spironolactone, 99%
Spironolactone, 99%
Thermo Scientific Chemicals
Spironolactone, 99%
CAS: 52-01-7 | C24H32O4S | 416.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR207460010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSpironolactone
Name Note99%
CAS52-01-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Suspected of causing cancer.
May damage fertility.
May cause damage to organs through prolonged or repeated exposure.