Polyphosphoric acid, pure, >84% phosphate (as P2O5), Thermo Scientific Chemicals
Polyphosphoric acid, pure, &gt;84% phosphate (as P<sub>2</sub>O<sub>5</sub>), Thermo Scientific Chemicals
Polyphosphoric acid, pure, &gt;84% phosphate (as P<sub>2</sub>O<sub>5</sub>), Thermo Scientific Chemicals
Polyphosphoric acid, pure, &gt;84% phosphate (as P<sub>2</sub>O<sub>5</sub>), Thermo Scientific Chemicals
Polyphosphoric acid, pure, &gt;84% phosphate (as P<sub>2</sub>O<sub>5</sub>), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polyphosphoric acid, pure, >84% phosphate (as P2O5), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number ACR196950025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS551-92-8
IUPAC Name1,2-dimethyl-5-nitro-1H-imidazole
Molecular FormulaC5H7N3O2
InChI KeyIBXPYPUJPLLOIN-UHFFFAOYSA-N
SMILESCN1C(C)=NC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)133.5-142.5?C