Tin(II) chloride dihydrate, 97%, Thermo Scientific Chemicals
Tin(II) chloride dihydrate, 97%, Thermo Scientific Chemicals
Tin(II) chloride dihydrate, 97%, Thermo Scientific Chemicals
Tin(II) chloride dihydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tin(II) chloride dihydrate, 97%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ACR196805000500 g
Catalog number ACR196805000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point652°C
Chemical Name or MaterialTin(II) chloride dihydrate
Melting Point37°C to 38°C
Name Note97%
CAS10025-69-1