hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt, Thermo Scientific Chemicals
hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt, Thermo Scientific Chemicals
hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt, Thermo Scientific Chemicals
hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt, Thermo Scientific Chemicals
hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt, Thermo Scientific Chemicals
Thermo Scientific Chemicals
hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt, Thermo Scientific Chemicals
CAS: 26023-84-7 | Cl6H2Pt | 409.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR195370050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS21653-40-7
IUPAC Namehydrogen 5-[(diaminomethylidene)amino]-N-(4-nitrophenyl)-2-(phenylformamido)pentanamide chloride
Molecular FormulaC19H23ClN6O4
InChI KeyDEOKFPFLXFNAON-UHFFFAOYNA-N
SMILES[H+].[Cl-].NC(N)=NCCCC(NC(=O)C1=CC=CC=C1)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.5 %
Infrared spectrumConforms
Specific optical rotation+42° to +48° (20°C, 589 nm) (c=1 in DMF)
Appearance (Color)White to yellow