Antimony, 99+%, powder, 0.15 mm
Antimony, 99+%, powder, 0.15 mm
Antimony, 99+%, powder, 0.15 mm
Antimony, 99+%, powder, 0.15 mm
Thermo Scientific Chemicals
Antimony, 99+%, powder, 0.15 mm
CAS: 7440-36-0 | Sb | 121.76 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR19245010010 gGlass bottle
Catalog number ACR192450100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10025-69-1
IUPAC Nameλ²-tin(2+) dihydrate dichloride
Molecular FormulaCl2H4O2Sn
InChI KeyFWPIDFUJEMBDLS-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder, crystals and chunks
Titration Iodimetric98.0 to 103.0 %
Sulfate (SO4)Passes test (limit about 0.003%)
Solubility(in hydrochloric acid) Passes test