DL-10-Camphorsulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
CAS: 34850-66-3 | C10H15NaO4S | 254.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR189120050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-10-Camphorsulfonic acid, sodium salt
Name Note98%
CAS34850-66-3
Melting Point286°C to 288°C