3-Fluoropyridine, 99%, Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR188300010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or pellets
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White