Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
CAS: 144-62-7 | C2H2O4 | 90.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number ACR186430010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialOxalic acid
CAS144-62-7
Melting Point189.0°C to 191.0°C
Health Hazard 1Danger
ColorWhite
View more
Suitable for battery materials development.
RUO – Research Use Only