Formamide, 99.5+%, extra pure, Thermo Scientific Chemicals
Formamide, 99.5+%, extra pure, Thermo Scientific Chemicals
Formamide, 99.5+%, extra pure, Thermo Scientific Chemicals
Formamide, 99.5+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formamide, 99.5+%, extra pure, Thermo Scientific Chemicals
CAS: 75-12-7 | CH3NO | 45.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1810900101 LGlass Bottle
Catalog number ACR181090010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS123-91-1
IUPAC Name1,4-dioxane
Molecular FormulaC4H8O2
InChI KeyRYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILESC1COCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤0.05%
StabilizerIf present, report quantity and identity
Appearance (Color)Colorless to pale yellow
FormLiquid
Color (APHA)≤20