Barbituric acid, 99+%, Thermo Scientific Chemicals
Barbituric acid, 99+%, Thermo Scientific Chemicals
Barbituric acid, 99+%, Thermo Scientific Chemicals
Barbituric acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Barbituric acid, 99+%, Thermo Scientific Chemicals
Barbituric acid, CAS # 67-52-7, (synonym: malonylurea) is the parent compound used in the production of barbiturates that act as sedative drugs. It is also used extensively in the manufacturing of plastics, textiles, and polymers.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ACR180925000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS142-71-2
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>97.5%
Appearance (Color)Blue-green
Elemental AnalysisCl:<200ppm
pH5.0-5.3
Formpowder
View more
General description
• Barbituric acid is a white, odorless, water-soluble crystalline powder
• It is a strong acid in an aqueous medium
Applications
• Barbituric acid uses the Knoevenegal condensation reaction to be converted into barbiturate drugs that acts as central nervous system depressants
• It is an active ingredient in the synthesis of Vitamin B12
• It is also used in electrochemical oxidation of iodine using cyclic voltammetry and controlled potential coulometry
RUO – Research Use Only
General References:
  1. Ernst, B. J.; Clark, G. F.; Grundmann, O. The Physicochemical and Pharmacokinetic Relationships of Barbiturates - From the Past to the Future. Curr Pharm Des 2015, 21(25), 3681-91.
  2. Lingens, B.; Schild, T. A.; Vogler, B.; and Renz, B. Biosynthesis of vitamin B12. Transformation of riboflavin 2H-labeled in the 1'R position of 1'S position into 5,6-dimethylbenzimidazole Eur J Biochem
  3. 1992, 207(3):981-5.
  4. Nematollahi, D.; Hesari, M. Electrochemical study of iodide in the presence of barbituric acid. Application to coulometric titration of barbituric acid. Microchemical Journal 2001, 70(1), 7-11.