Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
250 g
Packaging:
Glass Bottle
Catalog number ACR180200010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Boiling Point218.0°C
Chemical Name or MaterialNaphthalene
Melting Point79.0°C to 82.0°C
Name Note99+%
CAS91-20-3
View more
For fluorescence/luminescence spectroscopy
RUO – Research Use Only