1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals
1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals
1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals
1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals
CAS: 107-08-4 | C3H7I | 169.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Catalog number ACR174052500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS86-65-7
IUPAC Name7-aminonaphthalene-1,3-disulfonic acid
Molecular FormulaC10H9NO6S2
InChI KeyCMOLPZZVECHXKN-UHFFFAOYSA-N
SMILESNC1=CC=C2C=C(C=C(C2=C1)S(O)(=O)=O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to green-beige to light brown
Infrared spectrumConforms
Appearance (Form)Powder or needles
Titration with NaOH>=60 % (As is)