Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
CAS: 375-22-4 | C4HF7O2 | 214.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR172801000100 gGlass bottle
Catalog number ACR172801000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2905-56-8
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms to structure (U.S. specification)
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
FormLiquid
CommentSpecification differs for U.S. and non-U.S. material where indicated