Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic drum
Catalog number ACR171530010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS1448-87-9
IUPAC Name2-chloroquinoxaline
Molecular FormulaC8H5ClN2
InChI KeyBYHVGQHIAFURIL-UHFFFAOYSA-N
SMILESClC1=CN=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or solid
HPLC>=96.0 %
Appearance (Color)White to grey to brown
Infrared spectrumConforms
Melting point47°C to 51°C
Filter aid
RUO – Research Use Only