8-Pentadecanone, 98%
8-Pentadecanone, 98%
8-Pentadecanone, 98%
8-Pentadecanone, 98%
Thermo Scientific Chemicals
8-Pentadecanone, 98%
CAS: 818-23-5 | C15H30O | 226.40 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR16979025025 gGlass bottle
Catalog number ACR169790250
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10043-35-3
IUPAC Nameboric acid
Molecular FormulaBH3O3
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILESOB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<3 ppm
Titration with NaOH>=99.4 %
Phosphate (PO4)=<5 ppm
Magnesium (Mg)=<5 ppm
Arsenic (As)=<0.5 ppm