Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number ACR168880050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS618-87-1
IUPAC Name3,5-dinitroaniline
Molecular FormulaC6H5N3O4
InChI KeyMPBZUKLDHPOCLS-UHFFFAOYSA-N
SMILESNC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point159°C to 163°C
Appearance (Color)Yellow to light brown
Appearance (Form)Powder
HPLC>=97.5 %