6-Chloronicotinic acid, 99%, Thermo Scientific Chemicals
6-Chloronicotinic acid, 99%, Thermo Scientific Chemicals
6-Chloronicotinic acid, 99%, Thermo Scientific Chemicals
6-Chloronicotinic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
6-Chloronicotinic acid, 99%, Thermo Scientific Chemicals
CAS: 5326-23-8 | C6H4ClNO2 | 157.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR168691000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material6-Chloronicotinic acid
Name Note99%
CAS5326-23-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning