Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
CAS: 124-41-4 | CH3NaO | 54.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
10 L
25 L
Catalog number ACR168600250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS104-83-6
IUPAC Name1-chloro-4-(chloromethyl)benzene
Molecular FormulaC7H6Cl2
InChI KeyJQZAEUFPPSRDOP-UHFFFAOYSA-N
SMILESClCC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or clear colorless to pale yellow
Refractive Index1.5555-1.5595 @ 20?C
FormFused solid or liquid as melt
Assay (GC)≥98.0%