n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals
n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals
n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals
n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals
CAS: 123-86-4 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number ACR167680010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7791-20-0
IUPAC Namenickel(2+) hexahydrate dichloride
Molecular FormulaCl2H12NiO6
InChI KeyLAIZPRYFQUWUBN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Ni++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Appearance (Color)Green
Water Content (Karl Fischer Titration)41-49.31% (5-7 waters)
For UV/VIS spectroscopy
RUO – Research Use Only