Triphenylphosphine Sulfide, 99+%
Triphenylphosphine Sulfide, 99+%
Triphenylphosphine Sulfide, 99+%
Triphenylphosphine Sulfide, 99+%
Thermo Scientific Chemicals
Triphenylphosphine Sulfide, 99+%
CAS: 3878-45-3 | C18H15PS | 294.35 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR15903010010 gGlass bottle
Catalog number ACR159030100
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS122-39-4
IUPAC NameN-phenylaniline
Molecular FormulaC12H11N
InChI KeyDMBHHRLKUKUOEG-UHFFFAOYSA-N
SMILESN(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Sensitivity(to nitrate) Passes test
Appearance (Color)White to off-white
Sulfated ash=<0.05 %
Appearance (Form)Crystalline powder or crystals or flakes