Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number ACR158902500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3891-07-4
IUPAC Name2-(2-hydroxyethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC10H9NO3
InChI KeyMWFLUYFYHANMCM-UHFFFAOYSA-N
SMILESOCCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
HPLC>=98.5 % (on dry substance)
Appearance (Form)Crystalline powder
Melting point126°C to 129°C
Lewis acid
RUO – Research Use Only