Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number ACR158900500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBoron trifluoride dimethanol complex
Name Note50-52 wt% BF3
CAS2802-68-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.<br/
View more
Lewis acid
RUO – Research Use Only