Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Packaging:
Glass bottle
Catalog number ACR158900010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS51-20-7
IUPAC Name5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC4H3BrN2O2
InChI KeyLQLQRFGHAALLLE-UHFFFAOYSA-N
SMILESBrC1=CNC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Assay (HPLC)≥98.0%
FormPowder
Lewis acid
RUO – Research Use Only