2'-Acetonaphthone, 99%, Thermo Scientific Chemicals
2'-Acetonaphthone, 99%, Thermo Scientific Chemicals
2'-Acetonaphthone, 99%, Thermo Scientific Chemicals
2'-Acetonaphthone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2'-Acetonaphthone, 99%, Thermo Scientific Chemicals
CAS: 93-08-3 | C12H10O | 170.21 g/mol
Have Questions?
Catalog NumberQuantity
ACR158881000100 g
Catalog number ACR158881000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS123-08-0
IUPAC Name4-hydroxybenzaldehyde
Molecular FormulaC7H6O2
InChI KeyRGHHSNMVTDWUBI-UHFFFAOYSA-N
SMILESOC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point114°C to 118°C
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Yellow to light brown
Appearance (Form)Crystalline powder