β-D-Galactose Pentaacetate ≥99%
β-D-Galactose Pentaacetate ≥99%
β-D-Galactose Pentaacetate ≥99%
β-D-Galactose Pentaacetate ≥99%
Thermo Scientific Chemicals
β-D-Galactose Pentaacetate ≥99%
CAS: 4163-60-4 | C16H22O11 | 390.341 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR158611000100 gPlastic bottle
Catalog number ACR158611000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Materialβ-D-Galactose pentaacetate
Name Note99+%
CAS4163-60-4
CAS Min %99.0
CAS Max %100.0