2-Thiouracil, 99%, Thermo Scientific Chemicals
2-Thiouracil, 99%, Thermo Scientific Chemicals
2-Thiouracil, 99%, Thermo Scientific Chemicals
2-Thiouracil, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Thiouracil, 99%, Thermo Scientific Chemicals
2-Thiouracil, Thiouracil or 4-Hydroxy-2-mercaptopyrimidine, CAS # 141-90-2, is the modified analog of the nucleotide base, uracil.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
View all
Glass bottle
Catalog number ACR151840050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Thiouracil
CAS141-90-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
General description
• 2-Thiouracil (Thiouracil or 4-Hydroxy-2-mercaptopyrimidine) is a nucleotide analog with a C-2 thioxo substitution
• 2-Thiouracil is a melanocyte suppressor and thyroid inhibitor
Applications
• 2-Thiouracil can be used to inhibit nitric oxide synthase and suppresses nitric oxide signaling in vitro
• 2-Thiouracil can be used for labeling of RNA in vitro
• 2-Thiouracil is intended for research applications only
RUO – Research Use Only
General References:
  1. Yang, W.; Shen, Z.; Wen, S.; Wang, W.; Hu, M. Mechanisms of multiple neurotransmitters in the effects of Lycopene on brain injury induced by Hyperlipidemia. Lipids Health Dis. 2018, 17(1), 13. Erratum in: Lipids Health Dis. 2018, 17 (1), 65.
  2. Wauters, J.; Van, Meulebroek L.; Fichant, E.; Delahaut, P.; Vanhaecke, L. Discrimination between Synthetically Administered and Endogenous Thiouracil Based on Monitoring of Urine, Muscle, and Thyroid Tissue: An in Vivo Study in Young and Adult Bovines. J Agric Food Chem. 2017, 65(30), 6231-6239.